Composite Lint-Free Wipe, Medium Size, MicroCare
Supplier: MicroCare
MCC-W88
76020-246CS 923.8 USD
76020-246
Composite Lint-Free Wipe, Medium Size, MicroCare
Wipes Disposable Wipes and Towels Wipes for General Applications
This wipe offers great cleaning at low cost.
  • Heavy economy wipe
  • Great cleaning at low cost
  • Five layers of material in each wipe
  • Class 100 clean room packaging
Great cleaning at low cost. Heavy wipe: 4.0 oz. with five layers of material in each wipe. Resists damage from abrasion.
It is made of five (5) layers of different materials, which produces a wipe that is very absorbent, soft and fluffy. The wipe is 'apertured' (punched with microscopic holes) to enhance absorbency. The wipe is very strong and resists abrasion, so it’s an excellent choice when hard scrubbing is required.
Packaging: 100 sheets/bag, 12 bags/case.