General Purpose Wipe, Premium Grade, Medium Size, MicroCare
Supplier: MicroCare
MCC-W66DF
76020-243CS 1139.53 USD
76020-243
General Purpose Wipe, Premium Grade, Medium Size, MicroCare
Wipes Disposable Wipes and Towels Wipes for General Applications
This is the premier benchtop wipe sized for the rework bench.
  • Premium benchtop wipe
  • Using the famous 'FP' fabric only from MicroCare for better cleaning
  • Cleaner, stronger, better absorbency than competing wipes
  • Sized for the rework bench
Very strong, no cellulose, no polyster and no glues, so less linting and better cleaning. Cleaner, stronger, more absorbtion than competing wipes.
Convenient package size. 50% more absorption,  and 80% less linting than the best-selling benchmark, the W66 Lint-Free Wipe. Medium weight, superior purity. While it’s the optimal size for use on the rework bench, it’s probably too small for stencil cleaning. Smooth, white, medium absorbency, standard packaging.
Packaging: 50 sheets/bag, 25 bags/case.