SMT Stencil Wipe, Economy Grade, Large, MicroCare
Supplier: MicroCare
MCC-W11
76020-235CS 191.9 USD
76020-235
SMT Stencil Wipe, Economy Grade, Large, MicroCare
Wipes Disposable Wipes and Towels Wipes for General Applications
This is a good quality stencil wipe that offers excellent cleaning at an economical price.
  • SMT stencil wipe, economy quality with large size
  • Smooth, white, medium absorbency
  • Optimal size for cleaning large devices, like SMT stencil cleaning
It’s a large with, about 8 inches by 11", so it delivers lots of scrubbing. It is made from hydroentangled polyester and cellulose, without any glues or binders for better cleaning. The material is lighter than other choices, so it’s a better choice for less heavily contaminated surfaces. This wipe is packaged with the MicroCare reflow oven cleaner (#MCC-ROC) and the MicroCare aqueous stencil cleaner (#MCC-BGA).
Packaging: 100 sheets/bag, 6 bags/case.