FastDNA Spin Kit for Soil, 50 ml tubes, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
116560600
76020-016KT 353.45 USD
76020-016
FastDNA Spin Kit for Soil, 50 ml tubes, MP Biomedicals
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The FastDNA™-50 mL SPIN Kit for Soil is designed to efficiently isolate bacterial, fungi, plant and animal genomic DNA from soil and environmental samples.
  • Used for Soil, Feces, Environmental Water, Wastewater, and Sludge
  • Sample volume approximately 200 µL to 50 mL; up to approximately 25 g
  • DNA Enrichment
  • DNase and RNase Free
The FastDNA Spin Kit for Soil is designed to isolate bacterial, fungal, plant, and animal genomic DNA from soil and other environmental samples. Soil is processed by a FastPrep® instrument and Lysing Matrix E tubes designed to efficiently lyse all microorganisms. The released DNA is purified by a silica-based spin filter method and is suitable for PCR analysis and other downstream applications. Complete lysis of all organisms, including historically difficult sources such as eubacterial spores and endospores, gram positive bacteria, yeast, algae, nematodes and fungi is achieved within seconds using a FastPrep instrument and Lysing Matrix E tubes.
Packaging: Kit contains 10 x 50 mL Garnet Lysing Matrix Tubes, 60 mL PLS Solution, 15 mL MT Buffer, 350 mL Sodium Phosphate Buffer, 25 mL PPS for Soil, 120 mL Binding Matrix, 120 mL GTC Wash Buffer, 12 mL SEWS-M, 60 mL TES, 200 µL BBS Gel Loading Dye 200, 10 x 50 mL SPIN Filter Tubes