Ethidium Bromide Greenbag™ Disposal Kit, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
112350200
76020-002EA 341.65 USD
76020-002
Ethidium Bromide Greenbag™ Disposal Kit, MP Biomedicals
Electrophoresis Reagents Stain Removal Reagents
Each EtBr Green BagTM contains a standard amount of activated material that will safely and easily concentrate Ethidium Bromide from large liquid quantities
One kit has the capacity to remove 500 mg of ethidium bromide from solutions (10 mg EtBr/bag).
The bags containing the biohazardous material can then be disposed of in a proper container.