dNTPs Mix 5 mm, 10 mm and 25 mm, MP Biomedicals
Supplier: 0000012207
11NTPMX055 11NTPMX105 11NTPMX250 11NTPMX255 11NTPMX100 11NTPMX050
75857-004EA 795.29 USD
75857-004 75857-006 75857-008 75857-010 76019-992 76020-304
dNTPs Mix 5 mm, 10 mm and 25 mm, MP Biomedicals
Nucleic Acid Reagents Nucleotides
Ready to use solutions of dATP, dCTP, dGTP, dTTP.
  • Improve PCR efficiency
  • Lower denaturation temperatures
  • Increase template lengths
The quality of nucleotides used in a PCR can significantly influence the result of the reaction
dNTP mix contains 5ųmol of each dATP, dCTP, dGTP, dTTP in an aqueous solution nuetralized to pH 7.0 at 5mM.
Application: Nucleic Acid Sequencing, Nucleic Acid Labeling, Nucleic Acid Amplification, PCR, high fidelity and long PCR, LAMP-PCR, cDNA synthesis, RT-PCR, DNA Labeling, DNA sequencing
Our manufacturing process guarantees that the dNTPs are highly purified (>99%). Contaminants can often hinder PCR and result in disappointing product yield. Achieve sensitive and consistent results with our specially prepared deoxynucleotides. Also suitable for use in cDNA synthesis, DNA sequencing and labeling procedures.