GeneClean® Turbo Kit, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
111102400
76019-990EA 268.71 USD
76019-990
GeneClean® Turbo Kit, MP Biomedicals
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
GeneClean® Turbo kit is used for purification of DNA fragments of sizes 0.1 to 300 kb from agarose gels, PCR reactions and other enzymatic solutions. Purified DNA can be used in enzymatic reactions, conventional or automated sequencing, cloning and microarray analysis.
  • Isolate DNA from TAE or TBE buffered agarose gels, PCR reactions, and other enzymatic solutions
  • Save time with patented Turbo fi lter design
  • Process with either vacuum or centrifugation
  • Purify larger fragments due to less manipulation of bound DNA
  • No carryover of matrix into DNA solution
  • Fastest, most flexible kit
GeneClean® technology simplifies the process of purifying DNA into three easy steps: bind, wash, and elute. Ethanol precipitation is never required, and purified DNA is immediately ready for a wide variety of downstream applications.
GeneClean® Turbo Kit is a flexible kit designed for purification of DNA fragments of sizes 0.1 kb to 300 kb from TAE or TBE buffered agarose gels, PCR reactions and other enzymatic solutions. It purifies larger fragments due to less manipulation of bound DNA. Process with either vacuum or centrifugation and save time with patented turbo filter design.
Isolate DNA from: TAE or TBE buffered agarose gels, enzymatic reactions, buffered solutions (desalting), PCR products from templates, labeling reactions.
Application includes: Nucleic Acid Isolation and Purification.
All GeneClean® Turbo Kits are shipped ambiently. Store all reagents at room temperature (15 - 30 °C).