Access Key Options for Ultra-Low Temperature Freezers, Thermo Scientific
Supplier: Thermo Fisher Scientific
FAC34567LE
76019-738EA 1460 USD
76019-738
Access Key Options for Ultra-Low Temperature Freezers, Thermo Scientific
Security Locks
Record who opened the ultra low freezer door, when it was opened and how long it was open with access key options.
Ordering information: Field-Installed access key option includes card access with five key cards. All orders must accompany a TSX, Revco RLE, Forma 89000, TLE or HERAfreeze HLE Series ultra-low freezer. No stand-alone orders.