Stand for Model BDC250, Caframo
Supplier: Caframo
A215
76019-364EA 239.96 USD
76019-364
Stand for Model BDC250, Caframo
Stirrers Stirring Tools
Portable support stand for Caframo’s BDC250 Petite Stirrer (97001-138). The base is coated for protection from corrosion and chemical spills. Rod diameter is 10 mm (3/8").
  • Base is mild steel coated with chemical resistant epoxy paint (chemical and corrosion resistant – durable and easy to clean)
  • Small footprint – Excellent stability within limited bench space
  • Stand weighs less than 2.2 lbs. – Easily portable
Support stand for Caframo’s BDC250 Petite Stirrer (97001-138). Conveniently portable and easy to assemble for changing work stations or field work. The mild steel base is coated for protection from corrosion and chemical spills.
Ordering information: Included as part of BDC250 stirrer package.