Impeller Set (Shaft and Blade Kit) for Model BDC250, Caframo
Supplier: Caframo
S100
76019-362EA 158.6 USD
76019-362
Impeller Set (Shaft and Blade Kit) for Model BDC250, Caframo
Stirrers Stirring Blades
Impeller set contains two blades and shaft. Includes axial and radial 0.4×1.1" blades 73/8×0.24" shaft and hardware 304 stainless steel.
  • Electropolished 316L stainless steel – Chemical and corrosion resistant
  • Axial and radial blades included – Choice of two types of flow
  • Set screw compatible with standard tools – Blades easily interchangeable
Shaft and impeller blades to fit collet of BDC250 (97001-138) stirrer, all electropolished 316L stainless steel. Includes one radial blade and one axial blade and interchangeable set screw.
Ordering information: This kit is included when a BDC250 is purchased.