Stand Rods, Hollow, 1" Stainless Steel (304), Caframo
Supplier: Caframo
ROD48 ROD28 ROD38
52428-766EA 225.87 USD
52428-766 76019-350 76019-352
Stand Rods, Hollow, 1" Stainless Steel (304), Caframo
Solid Rods
Select the stand support rod that fits best with the height of your mixing requirements.
  • Made of 304 stainless steel – chemically inert and corrosion resistant
  • Compatible with A110 stand base (52428-764)
  • Flexible to customize a stand for taller vessels and chemical reaction systems
  • Install using base hex key – easy to adapt to apparatus needs
Delivery information: Supplied with plastic (LDPE-low density polyethylene) cap.