PCR Tube, Forensic DNA Grade, Eppendorf
Supplier: Eppendorf
0030124707
76019-010CS 128.08 USD
76019-010
PCR Tube, Forensic DNA Grade, Eppendorf
Tubes PCR Tubes
The Forensic DNA Grade product line encompasses consumables for DNA extraction, sample processing and PCR setup as well as for sample storage.
  • Positive air pressure prevents intrusion of dust
  • Strict adherence to cleaning procedures and protocols minimize the risk of contamination
A high degree of automation to minimize direct contact between staff and product.
Certifications: Certified purity grade according to ISO 18385.