Ezwaste® Replacement Filter ¹/₄" Elbow Fittings, Foxx Life Sciences
Supplier: VWR International
330-0943-OEM
76018-664PK 135.6 USD
76018-664
Ezwaste® Replacement Filter ¹/₄" Elbow Fittings, Foxx Life Sciences
Filters Liquid Process Filters
Replacement fittings for EZwaste® solvent waste system's fittings.
  • MNPT: ¹/₄"
  • Elbow filter fittings: 90°
  • Closed loop design which protects against leaks and spills
  • Improved lab safety
Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers and provides an easy mechanism of disposal for vapor wastes.