Mini Cuvette Stirrers, FireflySci
Supplier: FIREFLYSCI, INC.
P72
76016-690EA 39.76 USD
76016-690
Mini Cuvette Stirrers, FireflySci
Stirrers Magnetic Stirring Bars
Mini Spectrophotometer Precision Cell Stirrer: This PTFE magnetic centrifugal stirrer fits into cuvettes with smaller apertures and volumes and provides rapid vertical and horizontal mixing with a minimum of vortexing when placed on a magnetic stirring machine.
  • Dimensions: 7×2 mm
  • For use with narrow-necked and semi-micro cuvettes
Centrifugal pumping action generated by the cross channels in the upper face mixers without aeration. Photometric measurements can proceed uninterrupted while reagents are added.