Circular Dichroism Disc Mount (For Type 42 Cuvette), A26
Supplier: FIREFLYSCI, INC.
A26
76016-666EA 258.62 USD
76016-666
Circular Dichroism Disc Mount (For Type 42 Cuvette), A26
Cuvettes Spectrophotometer Cuvettes
Cuvette mount for Type 42 Circular Dichroism Disc Cells.
  • This mount fastens the two sides of the disc together
  • Compatible with Type 42 cuvettes
The A26 Cuvette mount is made from high-quality stainless steel. The mount works by applying pressure to the top release clamp, which opens the sides for insertion of the cuvette.