Cuvette Mount (For Type 507 and 607 Cuvettes), A24
Supplier: FIREFLYSCI, INC.
A24
76016-662EA 300.48 USD
76016-662
Cuvette Mount (For Type 507 and 607 Cuvettes), A24
Racks Cuvette Racks
Cell mount for Type 507 and Type 607 micro cells.
  • Anodized aluminum construction
  • External dimensions: 12.5L×12.5W×43H mm
Mount enables Type 507 and Type 607 cuvettes to fit in a standard 10 mm cuvette holder.