Cuvette Mount (For Type 4 Cuvettes), A23
Supplier: FIREFLYSCI, INC.
A23
76016-660EA 300.48 USD
76016-660
Cuvette Mount (For Type 4 Cuvettes), A23
Racks Cuvette Racks
Cell mount suitable for use with Type 4 cuvettes.
  • Anodized aluminum construction
  • External dimensions: 12.5L×12.5W×40H mm
The A23 cuvette mount enables a Type 4 cuvette to fit in a standard 12.5×12.5 mm cuvette holder.