Hardshell Plastic Cuvette Cases, Type P
Supplier: FIREFLYSCI, INC.
CASEP4
76016-134EA 10.64 USD
76016-134 76016-136 76016-652
Hardshell Plastic Cuvette Cases, Type P
Cuvettes Spectrophotometer Cuvettes
Cuvette cases hold 4 cells up to 10 mm L/P each, or two 20 mm L/P cells, or one 50 mm L/P cell.
  • Black opaque finish