Dye Laser Cell, FireFlySci
Supplier: FIREFLYSCI, INC.
2267A
76016-570EA 1129.31 USD
76016-570
Dye Laser Cell, FireFlySci
Cuvettes Spectrophotometer Cuvettes
This Dye Laser Cell is a standard 20 mm dye laser cell.
  • Highly accurate results, only need to test sample once
  • Made by hand, ensuring high quality optical polishing with no defects
  • Fire-fused to ensure that cuvettes are robust and durable
  • Available in UV Quartz and 20 mm lightpath
  • Four windows and base optically polished
  • High degree of parallelism for dye laser measurements
Each cell goes through a vigorous inspection process before leaving the factory.
Firefly cuvettes are made with high quality quartz and glass material available. Good transmission values.