Micro Flow Through Cell, Type 513M, Firefly Inc.
Supplier: FIREFLYSCI, INC.
513MUV10
76016-424EA 937.25 USD
76016-424
Micro Flow Through Cell, Type 513M, Firefly Inc.
Cuvettes Spectrophotometer Cuvettes
The micro flow through cell has PTFE inlet/outlet tubes with 4 mm outer diameter and 2 mm internal diameter.
  • Highly accurate results
  • Two windows polished
  • Excellent transmission values
  • Fire-fused to ensure that cuvettes are robust and durable
  • Contains a z-dimension of 8.5 mm
High quality optical polishing with no defects, available in several different materials - UV Quartz and IR Quartz.
The height information is without tubes, chamber dimensions: 10L×4W mm.