Colorimeter Cell (Lightpath: 40 mm), Type 97
Supplier: FIREFLYSCI, INC.
97G40
76016-378EA 214.64 USD
76016-378
Colorimeter Cell (Lightpath: 40 mm), Type 97
Cuvettes Spectrophotometer Cuvettes
Type 97 Colorimeter cell.
  • Fire-fused to ensure that cuvettes are robust and durable
  • Available in Optical Glass
  • 40 mm lightpath available
Two windows are optically polished.