Light-Scattering Fluorescence Cuvette with Rounded Bottom, Type 75
Supplier: FIREFLYSCI, INC.
75UV
76016-328EA 280.51 USD
76016-328
Light-Scattering Fluorescence Cuvette with Rounded Bottom, Type 75
Cuvettes Spectrophotometer Cuvettes
Round bottom centrifuge fluorometer cell for light-scattering measurements.
  • Fire-fused to ensure that cuvettes are robust and durable
  • Available in UV quartz
All surfaces are clear and optically polished.