Standard Anaerobic Absorbance Cell with Glass Pouch, Firefly Sci
Supplier: FIREFLYSCI, INC.
28G10
76015-738EA 742.25 USD
76015-738
Standard Anaerobic Absorbance Cell with Glass Pouch, Firefly Sci
Cuvettes Spectrophotometer Cuvettes
The Type 28 absorbance cell contains an inlet tube and a glass pouch chamber for catching excess gas produced by anaerobic experiments.
  • Highly accurate results, only need to test sample once
  • Made by hand, ensuring high quality optical polishing with no defects
  • Each cell goes through a vigorous inspection process before leaving our factory
  • Fire-fused to ensure that cuvettes are robust and durable
  • Available in 10 mm lightpath
  • Two windows optically polished
  • Overall height with the pouch is approximately 125 mm
Firefly cuvettes are made with the best quality quartz and glass material available. Excellent transmission values.