Dye Laser Flow Through Cell, Firefly Sci
Supplier: FIREFLYSCI, INC.
0210UV17
76015-078EA 1042.71 USD
76015-078
Dye Laser Flow Through Cell, Firefly Sci
Cuvettes Spectrophotometer Cuvettes
Dye laser cells are manufactured with extreme surface flatness extending to the edges of the cell.
  • Highly accurate results, only need to test sample once
  • Made by hand, ensuring high quality optical polishing with no defects
  • Each cell goes through a rigorous inspection process before leaving our factory
  • Fire-fused to ensure that cuvettes are robust and durable
  • Four windows optically polished
  • Open top and open bottom flow through cell
  • Originally made for the Quanta Ray dye laser machine
Firefly cuvettes are made with the best quality quartz and glass material available. Excellent transmission values. Flatness is checked via interferometer.