Quanta-Ray Dye Laser Flow Through Cells, FireFlySci
Supplier: FIREFLYSCI, INC.
0005-0023-2
76015-076EA 1016.77 USD
76015-076
Quanta-Ray Dye Laser Flow Through Cells, FireFlySci
Cuvettes Spectrophotometer Cuvettes
Dye laser cells are manufactured with extreme surface flatness extending to the edges of the cell.
  • Highly accurate results, only need to test sample once
  • Made by hand, ensuring high quality optical polishing with no defects
  • Each cell goes through a rigorous inspection process before leaving our factory
  • Fire-fused to ensure that cuvettes are robust and durable
  • Available in UV Quartz
  • Available in dual 6×12 mm lightpath
  • Four windows optically polished
  • Open top and open bottom flow through cell
Flatness is checked via interferometer. Premium quality Firefly cuvettes are made with the best quality quartz and glass material available transmission values.