Phosphate Buffered Saline Solution, 1X, Sterile, BiotechGrade, Spectrum Chemicals
Supplier: SPECTRUM CHEMICAL MFG CORP
P3033-500ML
76014-824EA 103 USD
76014-824 76014-826
Phosphate Buffered Saline Solution, 1X, Sterile, BiotechGrade, Spectrum Chemicals
Buffers Biological Buffers
Phosphate Buffered Saline Solution, 1X, Sterile, BiotechGrade is abbreviated as PBS and is used in biological research as a buffer solution containing sodium chloride and sodium phosphate, as well as potassium phosphate and potassium chloride in some formulations.
  • Sterile
  • BiotechGrade
  • DNase: none detected
It is also used in cell container rinsing and substance dilution.