Maltitol solution NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (2S,3R,4R,5R)-4-(((2R,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexane-1,2,3,5,6-pentaol

M1051-200LTBL
76014-592EA 4836.85 USD
76014-592
Maltitol solution NF
Maltitol

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₁₂H₂₄O₁₁
MW: 344.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006600
CAS Number: 585-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR