L(+)-Glutamic acid

Supplier: Spectrum Chemical Mfg. Corp.

GL125-100GM GL125-1KG GL125-25KGBL
75872-582EA 77.75 USD
75872-582 75872-584 76014-516
L(+)-Glutamic acid
L(+)-Glutamic acid

L-Glutamic Acid is also known as glutamic acid. It is used in the food industry most usually as a flavor with an umami taste.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
Storage Temperature: Ambient
MDL Number: MFCD00002634
CAS Number: 56-86-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1cb2faefcc53483f0b3bfccd6c12b611e487241&vid=a462989da97f7186f34fef7de9cad6596c2589ee&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR