D-(+)-Glucosamine hydrochloride ≥98%

Supplier: Spectrum Chemical Mfg. Corp.

GL110-1KG GL110-25GM GL110-500GM GL110-5KG GL110-25KGBL GL110-100GM GL110-50KGBL
700008-502EA 727.65 USD
700008-502 700008-504 700008-508 700008-510 700008-506 700008-500 76014-410
D-(+)-Glucosamine hydrochloride ≥98%
D-(+)-Glucosamine hydrochloride

D-Glucosamine Hydrochloride is an amino sugar and plays an important role in the biochemical synthesis of glycosylated lipids and proteins. It is most commonly used as a non-vitamin dietary supplement.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₁₃NO₅
Storage Temperature: Ambient
MDL Number: MFCD00135831
CAS Number: 66-84-2

Order Now

Specification Test Results

Assay 98 % min.
Moisture max.1.5 %
Heavy Metals max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR