Choline chloride USP dietary supplement

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-hydroxy-N,N,N-trimethylethanaminium chloride

C2058-12KG C2058-2.5KG C2058-500GM
76014-324EA 1338.3 USD
76014-324 76014-344 76014-346
Choline chloride USP dietary supplement
Choline chloride

Choline Chloride, USP Dietary Supplement.


All Spectrum Chemical USP Dietary Supplement grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: HOH₂CCH₂N(CH₃)₃Cl
MW: 139.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011721
CAS Number: 67-48-1
Merck Index: 13,02226
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77b015e76f45084bc46da3249578a85652cd6802&vid=85c8a0a1b152b861b3bc3ec8fe25a70509221d85&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR