Fumaric acid FCC

Supplier: Spectrum Chemical Mfg. Corp.

FU105-12KG FU105-2.5KG FU105-500GM FU105-50KGBL
76014-328EA 1417.8 USD
76014-328 76014-330 76014-476 76014-478
Fumaric acid FCC
Fumaric acid

Fumaric Acid, FCC is used as an acidity regulator and coagulant food additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₄H₄O₄
Storage Temperature: Ambient
MDL Number: MFCD00002700
CAS Number: 110-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR