Anti-GTF3C4 Rabbit Polyclonal Antibody

Supplier: Prosci
56-142
76011-386EA 640.99 USD
76011-386
Anti-GTF3C4 Rabbit Polyclonal Antibody
Antibodies
Essential for RNA polymerase III to make a number of small nuclear and cytoplasmic RNAs, including 5S RNA, tRNA, and adenovirus-associated (VA) RNA of both cellular and viral origin. Has histone acetyltransferase activity (HAT) with unique specificity for free and nucleosomal H3. May cooperate with GTF3C5 in facilitating the recruitment of TFIIIB and RNA polymerase through direct interactions with BRF1, POLR3C and POLR3F. May be localized close to the A box.

For WB starting dilution is: 1:1000 For FACS starting dilution is: 1:10~50

Type: Primary
Antigen: GTF3C4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR