Anti-TNIP1 Rabbit Polyclonal Antibody

Supplier: Prosci
55-570
76009-276EA 640.99 USD
76009-276
Anti-TNIP1 Rabbit Polyclonal Antibody
Antibodies
Interacts with zinc finger protein A20/TNFAIP3 and inhibits TNF-induced NF-kappa-B-dependent gene expression by interfering with an RIP-or TRAF2-mediated transactivation signal (By similarity). Increases cell surface CD4(T4) antigen expression. Interacts with HIV-1 matrix protein and is packaged into virions and overexpression can inhibit viral replication. May regulate matrix nuclear localization, both nuclear import of PIC (Preintegration complex) and export of GAG polyprotein and viral genomic RNA during virion production.

For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:50~100

Type: Primary
Antigen: TNIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR