Disposable Sweatbands, OK-1® Safety & Ergonomics, OccuNomix
Supplier: Occunomix International, LLC
SBR100
76006-508PK 100.12 USD
76006-508
Disposable Sweatbands, OK-1® Safety & Ergonomics, OccuNomix
Disposable Garments
Traditional absorbent cellulose sweatband.
  • Anti-mold treated
  • Pre-moistened
  • Dip in cold water, squeeze out, and wear