Butane Microtorch, with Two Tips, Master Appliance
Supplier: Safety & Industrial Supplies
467-MT-76
76006-430EA 109.23 USD
76006-430
Butane Microtorch, with Two Tips, Master Appliance
Welding Supplies
Meets CPSC requirements.
  • Plastic body
  • 2500 °F temperature maximum
  • Adjustable flame; broad or pinpoint
Includes stand, solder tip, hot air tip, and removable base.
Certifications: CSA/US; Meets CPSC Requirements; TUV/GS.