EZ-Vac™ Vacuum Manifold, Zymo Research
Supplier: Zymo Research
S7000
76005-132EA 331.81 USD
76005-132
EZ-Vac™ Vacuum Manifold, Zymo Research
Manifolds
The EZ-Vac™ Vacuum Manifold features durable chemical-resistant construction and is capable of processing up to 20 samples simultaneously using vacuum pressure without the need for repeated centrifugation steps
  • Compatible with many spin-column based nucleic acid purification kits
  • Unique design accommodates large processing volumes easily
  • Simplifies the nucleic acid purification workflows
  • The One-Way Luer-lock Stopcocks are individually controlled, allowing users to process as little as one sample at a time
The EZ-Vac™ Vacuum Manifold consist of a linear, low-density polyethylene (LLDPE) manifold body, One-Way Luer-Lock Stopcocks made of a polycarbonate body and polypropylene handle, a polypropylene vacuum hose connector and a neoprene stopper.