Durx® 670 Polyester/Cellulose Nonwoven Flat Head Mop Replacement Covers, Berkshire
Supplier: Berkshire
DR670.0812.20
76003-524CS 252.08 USD
76003-524
Durx® 670 Polyester/Cellulose Nonwoven Flat Head Mop Replacement Covers, Berkshire
Mops
Durx® 670 flat mop replacement covers are hydro entangled 55% cellulose/45% polyester, and are designed to fit flat mop head frame (large and small sizes available). Flat mop replacement covers are made from polyester/cellulose nonwoven material.
  • Excellent combination of synthetic polyester strength and cleanliness, with the absorbent characteristics of cellulose
  • Smooth, highly absorbent, and durable
  • Low extractable levels and particle counts
  • Chemically compatible with common cleaning and disinfecting solutions
An economical alternative to knitted polyester replacement covers.