Screen-Well® Nephrotoxicity Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
LIB100-0100
76003-306EA 2972.43 USD
76003-306
Screen-Well® Nephrotoxicity Library, Enzo Life Sciences
Cell Culture Supplements and Additives
Essential standards for predictive toxicology screening.
The Screen-Well® Nephrotoxicity Library is a focused collection of 86 compounds with defined and diverse nephrotoxicity, including analgesic nephropathy, interstitial nephritis, glomerularopathy, proximal and distal tubulopathy, acute renal failure, chronic kidney disease, and more. A variety of structurally and mechanistically different compound classes are included, as well as nontoxic controls. Most of the compounds are dissolved in DMSO at 10mM unless otherwise stated in the documentation provided with the library. Selected compounds are dissolved in DI water. All compounds are provided at 100µL volumes in a 96-Well plate. This library is an essential cost-effective tool for predictive toxicology screening and assay development.
Ordering information: Contains 86 compounds with defined and diverse nephrotoxicity. A variety of structurally and mechanistically different compound classes, as well as nontoxic controls, are included.