PCR Decontamination Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT137-0100
76003-302EA 180.94 USD
76003-302
PCR Decontamination Kit, Enzo Life Sciences
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Rapid removal of contaminating DNA in PCR mastermixes, without reduction in PCR sensitivity.
  • Removes DNA contamination from PCR mastermixes
  • Improves target detection by reducing background
  • Does not effect PCR sensitivity
  • Optimized for PCR and probe based qPCR mixes
For the removal of contaminating DNA from primers and mastermixes prior to PCR.
The PCR decontamination kit uses a dsDNase to remove contaminating DNA from mastermixes. The double-strand specific property allows decontamination in the presence of primers and probe.The dsDNase is irreversibly inactivated by heating to 60°C in the presence of DTT, ensuring that DNA template added to the mastermix after inactivation will not be digested. The mastermix can be used immediately after decontamination to run PCR reactions.
AMPIGENE® Taq polymerases have been treated to remove host DNA and additional clean-up is not necessary. The PCR decontamination kit is recommended for decontamination of primers or competitor’s PCR mixes.
Ordering information: Contains Heat-labile dsDNase, DTT.