(R)-N-(2,3-Dihydroxypropoxy)-3,4-difluoro-2-(2-fluoro-4-iodophenylamino)benzamide ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: (R)-N-(2,3-Dihydroxypropoxy)-3,4-difluoro-2-((2-fluoro-4-iodophenyl)amino)benzamide, Mirdametinib, PD-0325901

CHM202-0010
76003-262EA 163.5 USD
76003-262
(R)-N-(2,3-Dihydroxypropoxy)-3,4-difluoro-2-(2-fluoro-4-iodophenylamino)benzamide ≥99% (by HPLC)
(R)-N-(2,3-Dihydroxypropoxy)-3,4-difluoro-2-(2-fluoro-4-iodophenylamino)benzamide

Selective MEK inhibitor


PD0325901 (PD325901) is selective and non ATP-competitive MEK inhibitor with an IC50 of 0.33 nM, roughly 500-fold more potent than CI-1040 on the phosphorylation of ERK1 and ERK2.

Formula: C₁₆H₁₄F₃IN₂O₄
MDL Number: MFCD08435926
CAS Number: 391210-10-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f88fe069c5295ed4afcc8ad143c3fa41985c62a5&vid=c94aee5029b16d3d329627008e5315062fe69ed5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR