Anti-IL7R Mouse Monoclonal Antibody [clone: R34-34]

Supplier: Enzo Life Sciences
ABS253-0100
76002-858EA 545 USD
76002-858
Anti-IL7R Mouse Monoclonal Antibody [clone: R34-34]
Antibodies
Recommended Applications: Flow Cytometry, IP, FUNC

Species reactivity: Human, Dog

Type: Primary
Antigen: IL7R
Clonality: Monoclonal
Clone: R34-34
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR