BIO-PROBE® Lambda Probe, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-40840
76002-526EA 415.29 USD
76002-526
BIO-PROBE® Lambda Probe, Enzo Life Sciences
Nucleic Acid Reagents Primers and Probes
Negative control probe for in situ hybridization.
The Lambda BIO-PROBE® labeled probe is prepared by nick translation of the lambda bacteriophage strain c1857ind1 Sam7. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).