Anti-CD80 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: mAb 104]

Supplier: Enzo Life Sciences
ABS151-0100
76002-436EA 774.99 USD
76002-436
Anti-CD80 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: mAb 104]
Antibodies
Recommended Applications: Flow Cytometry

Species reactivity: Human

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: mAb 104
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR