Rapamycin ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: (3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,27R,34aS)-9,10,12,13,14,21,22,23,24,25,26,27,32,33,34,34a-Hexadecahydro-9,27-dihydroxy-3-[(1R)-2-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethyl]-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-23,27-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine-1,5,11,28,29(4H,6H,31H)-pentone

BML-A275-0005 BML-A275-0025
89156-364EA 163.5 USD
89156-364 76002-362
Rapamycin ≥98% (by HPLC)
Rapamycin

Macrocyclic-triene antibiotic possessing potent immunosuppressant activity


  • Off-white or yellow solid


It forms a complex with FKBP12 that binds to an effector, thus inhibiting IL-2 and other growth promoting lymphokines. The effectors were identified as FRAP (FKBP12 rapamycin-associated protein) and RAFT1 (rapamycin and KFBP12 target). Rapamycin/FKBP complex does not inhibit FRAP PI 4-kinase activity, but does inhibit FRAP autophosphorylation. Rapamycin induces inhibition of p70s6k, p33cdk2 and p34cdc2. Selectively blocks signaling leading to the activation of p70/85 S6 kinase. Enhances apoptosis. Activator of autophagy both in vitro and in vivo.


Soluble in chloroform, DMSO (25 mg/mL) or methanol


Packaging: Shipping temperature: Ambient

Formula: C₅₁H₇₉NO₁₃
Storage Temperature: Freezer
MDL Number: MFCD00867594
CAS Number: 53123-88-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b47a70d6a13056b11b7542d50eef34075c75915&vid=4bf51ba62e5bcd1c0c5f619b7dcddca81022d3e3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR