SCREEN-WELL® Epigenetics Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-2836-0100 BML-2836-0500
76002-344EA 1540.17 USD
76002-344 76002-346
SCREEN-WELL® Epigenetics Library, Enzo Life Sciences
Cell Culture Supplements and Additives
Useful tool for chemical genomics, assay development, or other pharmacological applications in the study of epigenetic modifications of lysine and DNA methylation.
The Screen-Well® Epigenetics library contains 43 compounds with defined activity against enzymes which carry out epigenetic modification of lysine. It also includes DNA methylation inhibitors. Compounds are dissolved in DMSO at 10 mM and aliquoted into deep-well plates at 100 or 500 µL per well. The library is a useful tool for chemical genomics, assay development and other pharmacological applications. The library contains inhibitors of these important enzymes: HDACs, SIRTs, Lysine demethylases, HATs, Histone methyl transferases, DNA methyltransferases as well as SIRT activators. A variety of structurally and mechanistically different compound classes are included.
Ordering information: Contains 43 compounds supplied in DMSO at a concentration of 10 mM.