Anti-PTMA Mouse Monoclonal Antibody [clone: 1G8]

Supplier: Enzo Life Sciences
804-916-0100
76002-236EA 841.48 USD
76002-236
Anti-PTMA Mouse Monoclonal Antibody [clone: 1G8]
Antibodies
Recommended Applications: ELISA, WB

Species reactivity: Human

Type: Primary
Antigen: PTMA
Clonality: Monoclonal
Clone: 1G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR