Eicosapentaenoyl ethanolamide ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-FA047-0025
76002-110EA 267.05 USD
76002-110
Eicosapentaenoyl ethanolamide ≥98% (by TLC)
Eicosapentaenoyl ethanolamide

Synthetic.


Synthetic.

Has been shown to inhibit dietary-restriction-induced lifespan extension in wild-type and TOR pathway mutant nematodes. This data indicates that reduced N-acylethanolamide signaling mediates some of the effect of dietary-restriction-induced lifespan extension and that EPEA, specifically, acts as a metabolic signal that couples nutrient availability with growth and lifespan. EPEA has also been shown to have anti-proliferative effects and anti-inflammatory properties. Compound is subject to oxidation. Protect from air.


Has been shown to inhibit dietary-restriction-induced lifespan extension in wild-type and TOR pathway mutant nematodes. This data indicates that reduced N-acylethanolamide signaling mediates some of the effect of dietary-restriction-induced lifespan extension and that EPEA, specifically, acts as a metabolic signal that couples nutrient availability with growth and lifespan. EPEA has also been shown to have anti-proliferative effects and anti-inflammatory properties. Compound is subject to oxidation. Protect from air.

MW: 345.5 g/mol
CAS Number: 109001-03-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32e71d53e191839537f5d0f911f1dc8b12f5c4b7&vid=545828bb6d8596e90dc3eeac70ba8447648c0af0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR