Croton oil

Supplier: Enzo Life Sciences
Danger

Synonyms: Tiglium oil

350-089-KI225
76001-992EA 3799.74 USD
76001-992
Croton oil
Croton oil

Natural source of phorbol and phorbol esters.


Purified from Croton tiglium L.


Soluble in DMSO, 100% ethanol or acetone. Insoluble in cold water.

Storage Temperature: Refrigerator
MDL Number: MFCD00130880
CAS Number: 8001-28-3
Merck Index: 14,02598

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR