bpV(HOpic) ≥90% (by NMR)

Supplier: Enzo Life Sciences

270-206-M005 270-206-M025
89147-378EA 107.91 USD
89147-378 76001-966
bpV(HOpic) ≥90% (by NMR)
bpV(HOpic)

PTP inhibitor, PTEN Inhibitor


Protein phosphotyrosine phosphatase inhibitor.

Formula: K₂[VO(O₂)₂C₆H₄NO₃]
CAS Number: 722494-26-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d0ddc72541dcd66c9ddca48bd1ac9b9e0985770&vid=921091867343a654481e907cb1cb1a7c0e32a5de&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR