VT1000 S Vibrating Blade Microtome, Leica
Supplier: Leica Biosystems MS
14047235613 14041157009
76001-014EA 32883.58 USD
76001-014 76001-016 MSPP-140411570
VT1000 S Vibrating Blade Microtome, Leica
Microtomes and Microtome Assemblies Microtomes
The Leica VT1000 S vibrating blade microtome is the instrument of choice for high-quality sectioning requirements in neurophysiology, neuropathology experimental pathology, botany (roots and plants) and industry (foams).
  • Electronic adjustable cutting window
  • Minimum section thickness achievable with that instrument is around 15 µm
  • Amplitude 0.2; 0.4; 0.8; 1.0 mm
The Leica VT1000 S vibrating blade microtome has been designed for special applications associated with sectioning of fresh biological tissues. Due to its unique design, the Leica VT1000 S microtome is also the ideal instrument for sectioning various industrial and plant materials (foams, soft rubbers, various plant parts as for example roots).
Ordering information: Power cord is required and sold separately. For options, contact your Avantor representative.